Assay Details
| Gene Symbol: tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200176.1 | NP_956470.1 | 633 | 83 | ||
| GenBank mRNA | BC045460.1 | - | 633 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tmem38b| Gene Symbol: tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200176.1 | NP_956470.1 | 633 | 83 | ||
| GenBank mRNA | BC045460.1 | - | 633 | 83 | ||
Set Membership: |
|