Assay Details
| Gene Symbol: hebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200198.1 | NP_956492.1 | 4 - 5 | 532 | 94 | |
| GenBank mRNA | BC045936.1 | - | 4 - 5 | 532 | 94 | |
| HQ450691.1 | - | 4 - 5 | 94 | |||
More Information
Set Membership: |
|

hebp2
Genomic Map