Assay Details
| Gene Symbol: ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200316.1 | NP_956610.1 | 6 - 7 | 945 | 70 | |
| GenBank mRNA | BC051126.1 | - | 6 - 7 | 945 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ttc25| Gene Symbol: ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200316.1 | NP_956610.1 | 6 - 7 | 945 | 70 | |
| GenBank mRNA | BC051126.1 | - | 6 - 7 | 945 | 70 | |
Set Membership: |
|