Assay Details
| Gene Symbol: fbxo43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200431.2 | NP_956725.2 | 6 - 7 | 1447 | 92 | |
| XM_021466645.1 | XP_021322320.1 | 6 - 7 | 1459 | 92 | ||
| GenBank mRNA | BC097058.1 | - | 6 - 7 | 1451 | 92 | |
More Information
Set Membership: |
|

fbxo43
Genomic Map