Assay Details
| Gene Symbol: zeb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114551.2 | NP_001108023.2 | 2 - 3 | 263 | 65 | |
| NM_001135104.2 | NP_001128576.2 | 2 - 3 | 263 | 65 | ||
| XM_005167523.4 | XP_005167580.4 | 1 - 2 | 412 | 65 | ||
| XM_021478582.1 | XP_021334257.1 | 2 - 3 | 560 | 65 | ||
| GenBank mRNA | BC168514.1 | - | 2 - 3 | 243 | 65 | |
| EU379558.1 | - | 1 - 2 | 74 | 65 | ||
More Information
Set Membership: |
|

zeb2a
Genomic Map