Assay Details
Gene Symbol: dipk2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114926.1 | NP_001108398.1 | 1 - 2 | 758 | 68 | |
XM_005162483.4 | XP_005162540.1 | 2 - 3 | 1284 | 68 | ||
GenBank mRNA | BC155066.1 | - | 1 - 2 | 758 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
