Assay Details
| Gene Symbol: tmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115133.1 | NP_001108605.1 | 6 - 7 | 642 | 66 | |
| GenBank mRNA | BC056715.1 | - | 6 - 7 | 613 | 66 | |
More Information
Set Membership: |
|

tmx1
Genomic Map