Assay Details
| Gene Symbol: mrpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122621.1 | NP_001116093.1 | 3 - 4 | 214 | 58 | |
| GenBank mRNA | BC159118.1 | - | 3 - 4 | 214 | 58 | |
More Information
Set Membership: |
|

mrpl22
Genomic Map