Assay Details
| Gene Symbol: mboat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122944.1 | NP_001116416.1 | 2 - 3 | 350 | 111 | |
| XM_021476326.1 | XP_021332001.1 | 3 - 4 | 361 | 111 | ||
| GenBank mRNA | EU518498.1 | - | 2 - 3 | 350 | 111 | |
More Information
Set Membership: |
|

mboat4
Genomic Map