Assay Details
| Gene Symbol: tbcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212775.1 | NP_997940.1 | 2 - 3 | 338 | 83 | |
| GenBank mRNA | BC044438.1 | - | 2 - 3 | 338 | 83 | |
| BC153663.1 | - | 2 - 3 | 334 | 83 | ||
| BC164828.1 | - | 1 - 2 | 149 | 83 | ||
More Information
Set Membership: |
|

tbcb
Genomic Map