Assay Details
| Gene Symbol: TLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081796.1 | NP_001075265.1 | 2 | 2848 | 87 | |
| XM_023626043.1 | XP_023481811.1 | 2915 | 87 | |||
| XM_023626055.1 | XP_023481823.1 | 3530 | 87 | |||
| XM_023626061.1 | XP_023481829.1 | 5076 | 87 | |||
| XM_023626073.1 | XP_023481841.1 | 3439 | 87 | |||
| XM_023626080.1 | XP_023481848.1 | 4984 | 87 | |||
| GenBank mRNA | AY429602.1 | - | 2 | 2848 | 87 | |
| JL638254.1 | - | 1 | 2117 | 87 | ||
More Information
Set Membership: |
|

TLR2
Genomic Map