Assay Details
Gene Symbol: MIR182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032821.1 | 1 | 35 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear respiratory factor 1 | NRF1 | -11.2kb | Anti-sense | Chr. 4 - 84436433 - 84559996 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-96 | MIR96 | eca-mir-96 | 4.4kb | Sense | Chr. 4 - 84575663 - 84575740 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
