Assay Details
| Gene Symbol: MIRLET7E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032887.1 | 1 | 43 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-99b | MIR99B | eca-mir-99b | -108.0 | Sense | Chr. 10 - 21666034 - 21666103 on NCBI Build EquCab3.0 |
| Downstream | microRNA mir-125a | MIR125A | eca-mir-125a | 428.0 | Sense | Chr. 10 - 21666717 - 21666784 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIRLET7E
Genomic Map