Assay Details
Gene Symbol: MIR8976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_127982.1 | 1 | 69 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | inhibin subunit alpha | INHA | -30.4kb | Sense | Chr. 6 - 8836712 - 8839510 on NCBI Build EquCab3.0 | |
Downstream | solute carrier family 4 member 3 | SLC4A3 | 12.8kb | Sense | Chr. 6 - 8882804 - 8896482 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
