Assay Details
Gene Symbol: MIR9087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128110.1 | 1 | 80 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RAS guanyl releasing protein 4 | RASGRP4 | -73.9kb | Sense | Chr. 10 - 9615802 - 9629291 on NCBI Build EquCab3.0 | |
Downstream | mitogen-activated protein kinase kinase kinase kinase 1 | MAP4K1 | 35.9kb | Sense | Chr. 10 - 9739175 - 9756564 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
