Assay Details
| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256936.2 | NP_001243865.1 | 1 | 1922 | 83 | |
| XM_005609026.3 | XP_005609083.1 | 2068 | 83 | |||
| XM_005609027.3 | XP_005609084.1 | 2161 | 83 | |||
| XM_005609028.3 | XP_005609085.1 | 2157 | 83 | |||
| XM_005609029.3 | XP_005609086.1 | 2052 | 83 | |||
| XM_005609030.3 | XP_005609087.1 | 2016 | 83 | |||
| XM_005609031.3 | XP_005609088.2 | 3214 | 83 | |||
| XM_005609032.3 | XP_005609089.1 | 2093 | 83 | |||
| XM_014738884.2 | XP_014594370.1 | 2041 | 83 | |||
| XM_023638743.1 | XP_023494511.1 | 2045 | 83 | |||
| GenBank mRNA | JL637779.1 | - | 1 | 696 | 83 | |
More Information
Set Membership: |
|

FIGNL1
Genomic Map