Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257187.1 | NP_001244116.1 | 1 | 1425 | 67 | |
| XM_005599155.3 | XP_005599212.1 | 2227 | 67 | |||
| XM_005599157.3 | XP_005599214.1 | 2224 | 67 | |||
| XM_023616936.1 | XP_023472704.1 | 1921 | 67 | |||
| XM_023616937.1 | XP_023472705.1 | 2159 | 67 | |||
More Information
Set Membership: |
|

ANKRD34B
Genomic Map