Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309302.1 | NP_001296231.1 | 4 - 5 | 723 | 90 | |
| GenBank mRNA | JL616378.2 | - | 4 - 5 | 723 | 90 | |
| Gene Symbol: LOC111771317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632608.1 | XP_023488376.1 | 741 | 90 | ||
More Information
Set Membership: |
|

RRP15
Genomic Map