Assay Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618821.1 | XP_023474589.1 | 1240 | 74 | ||
| XM_023618822.1 | XP_023474590.1 | 1529 | 74 | |||
More Information
Set Membership: |
|

TRABD2A
Genomic Map