Assay Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731734.2 | XP_014587220.2 | 525 | 69 | ||
| XM_014731736.2 | XP_014587222.2 | 1185 | 69 | |||
| XM_014731737.2 | XP_014587223.2 | 1176 | 69 | |||
More Information
Set Membership: |
|

ZNF662
Genomic Map