Assay Details
| Gene Symbol: PCDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621665.1 | XP_023477433.1 | 4741 | 104 | ||
| XM_023621666.1 | XP_023477434.1 | 4449 | 104 | |||
More Information
Set Membership: |
|

PCDH8
Genomic Map