Assay Details
| Gene Symbol: PLAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601889.3 | XP_005601946.1 | 261 | 72 | ||
| XM_014732815.2 | XP_014588301.1 | 262 | 72 | |||
| XM_014732818.2 | XP_014588304.1 | 258 | 72 | |||
| XM_023623509.1 | XP_023479277.1 | 334 | 72 | |||
More Information
Set Membership: |
|

PLAAT1
Genomic Map