Assay Details
| Gene Symbol: LOC100063291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494568.5 | XP_001494618.3 | 1663 | 103 | ||
| XM_005603563.3 | XP_005603620.1 | 1051 | 103 | |||
| XM_014734711.2 | XP_014590197.2 | 1436 | 103 | |||
| XM_023624443.1 | XP_023480211.1 | 1518 | 103 | |||
| XM_023624444.1 | XP_023480212.1 | 1641 | 103 | |||
| XM_023624445.1 | XP_023480213.1 | 1385 | 103 | |||
More Information
Set Membership: |
|

LOC100063291
Genomic Map