Assay Details
| Gene Symbol: CLIC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493952.4 | XP_001494002.2 | 1876 | 85 | ||
| XM_023630089.1 | XP_023485857.1 | 2281 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLIC6| Gene Symbol: CLIC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493952.4 | XP_001494002.2 | 1876 | 85 | ||
| XM_023630089.1 | XP_023485857.1 | 2281 | 85 | |||
Set Membership: |
|