Assay Details
| Gene Symbol: CCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738298.2 | XP_014593784.1 | 516 | 93 | ||
| XM_023637042.1 | XP_023492810.1 | 273 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCL17| Gene Symbol: CCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738298.2 | XP_014593784.1 | 516 | 93 | ||
| XM_023637042.1 | XP_023492810.1 | 273 | 93 | |||
Set Membership: |
|