Assay Details
| Gene Symbol: ZNF425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639905.1 | XP_023495673.1 | 302 | 91 | ||
| XM_023639906.1 | XP_023495674.1 | 236 | 91 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF425| Gene Symbol: ZNF425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639905.1 | XP_023495673.1 | 302 | 91 | ||
| XM_023639906.1 | XP_023495674.1 | 236 | 91 | |||
Set Membership: |
|