Assay Details
| Gene Symbol: LRRC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649259.1 | XP_023505027.1 | 2330 | 70 | ||
| XM_023649260.1 | XP_023505028.1 | 2330 | 70 | |||
| XM_023649261.1 | XP_023505029.1 | 952 | 70 | |||
| XM_023649262.1 | XP_023505030.1 | 591 | 70 | |||
| XM_023649263.1 | XP_023505031.1 | 392 | 70 | |||
| XM_023649264.1 | XP_023505032.1 | 363 | 70 | |||
| XM_023649266.1 | XP_023505034.1 | 523 | 70 | |||
More Information
Set Membership: |
|

LRRC24
Genomic Map