Assay Details
| Gene Symbol: ARSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014728748.2 | XP_014584234.1 | 124 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014728748.2 transcripts.
Set Membership: |
|

ARSF
Genomic Map