Assay Details
| Gene Symbol: AKAP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614540.3 | XP_005614597.1 | 305 | 96 | ||
| XM_014729142.2 | XP_014584628.1 | 279 | 96 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AKAP14| Gene Symbol: AKAP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614540.3 | XP_005614597.1 | 305 | 96 | ||
| XM_014729142.2 | XP_014584628.1 | 279 | 96 | |||
Set Membership: |
|