Assay Details
| Gene Symbol: IL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596548.3 | XP_005596605.1 | 792 | 67 | ||
| XM_023650533.1 | XP_023506301.1 | 712 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596548.3 | XP_005596605.1 | 792 | 67 | ||
| XM_023650533.1 | XP_023506301.1 | 712 | 67 | |||
Set Membership: |
|