Assay Details
| Gene Symbol: ELFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655115.1 | XP_023510883.1 | 1548 | 102 | ||
| XM_023655116.1 | XP_023510884.1 | 1368 | 102 | |||
| XM_023655117.1 | XP_023510885.1 | 1246 | 102 | |||
More Information
Set Membership: |
|

ELFN1
Genomic Map