Assay Details
| Gene Symbol: AHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023635217.1 | XP_023490985.1 | 176 | 101 | ||
| XM_023635222.1 | XP_023490990.1 | 176 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023635217.1, XM_023635222.1 transcripts.
Set Membership: |
|

AHDC1
Genomic Map