Assay Details
| Gene Symbol: ZBTB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624665.1 | XP_023480433.1 | 607 | 86 | ||
| XM_023624666.1 | XP_023480434.1 | 554 | 86 | |||
| XM_023624667.1 | XP_023480435.1 | 1313 | 86 | |||
| XM_023624668.1 | XP_023480436.1 | 1305 | 86 | |||
More Information
Set Membership: |
|

ZBTB12
Genomic Map