Assay Details
| Gene Symbol: GPR63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501202.5 | XP_001501252.3 | 253 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001501202.5 transcripts.
Set Membership: |
|

GPR63
Genomic Map