Assay Details
| Gene Symbol: HS3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500813.6 | XP_001500863.2 | 508 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS3ST2| Gene Symbol: HS3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500813.6 | XP_001500863.2 | 508 | 96 | ||
Set Membership: |
|