Assay Details
| Gene Symbol: LRRC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601894.3 | XP_005601951.2 | 114 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005601894.3 transcripts.
Set Membership: |
|

LRRC15
Genomic Map