Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603650.3 | XP_005603707.1 | 1189 | 79 | ||
| XM_023624133.1 | XP_023479901.1 | 571 | 79 | |||
| XM_023624134.1 | XP_023479902.1 | 571 | 79 | |||
| XM_023624135.1 | XP_023479903.1 | 571 | 79 | |||
| GenBank mRNA | JL634877.1 | - | 1 - 2 | 130 | 79 | |
More Information
Set Membership: |
|

ZSCAN16
Genomic Map