Assay Details
| Gene Symbol: BBS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488843.4 | XP_001488893.2 | 208 | 85 | ||
| XM_005606489.3 | XP_005606546.2 | 208 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BBS10| Gene Symbol: BBS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488843.4 | XP_001488893.2 | 208 | 85 | ||
| XM_005606489.3 | XP_005606546.2 | 208 | 85 | |||
Set Membership: |
|