Assay Details
| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610123.3 | XP_005610180.2 | 827 | 70 | ||
| XM_014739846.2 | XP_014595332.1 | 805 | 70 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610123.3 | XP_005610180.2 | 827 | 70 | ||
| XM_014739846.2 | XP_014595332.1 | 805 | 70 | |||
Set Membership: |
|