Assay Details
| Gene Symbol: RAB39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501007.6 | XP_001501057.1 | 382 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB39A| Gene Symbol: RAB39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501007.6 | XP_001501057.1 | 382 | 87 | ||
Set Membership: |
|