Assay Details
| Gene Symbol: ZBTB33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614474.2 | XP_005614531.1 | 687 | 93 | ||
| XM_005614475.2 | XP_005614532.1 | 500 | 93 | |||
| XM_023633757.1 | XP_023489525.1 | 619 | 93 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005614474.2, XM_005614475.2, XM_023633757.1 transcripts.
Set Membership: |
|

ZBTB33
Genomic Map