Assay Details
| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917041.4 | XP_001917076.3 | 760 | 101 | ||
| XM_023641522.1 | XP_023497290.1 | 529 | 101 | |||
| XM_023641529.1 | XP_023497297.1 | 369 | 101 | |||
More Information
Set Membership: |
|

UBTD1
Genomic Map