Assay Details
| Gene Symbol: NGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496187.5 | XP_001496237.2 | 218 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001496187.5 transcripts.
Set Membership: |
|

NGF
Genomic Map