Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741399.2 | XP_014596885.1 | 1773 | 84 | ||
| XM_014741473.2 | XP_014596959.1 | 1311 | 84 | |||
| XM_014741528.2 | XP_014597014.1 | 1586 | 84 | |||
| XM_014741583.2 | XP_014597069.1 | 503 | 84 | |||
| XM_014741589.2 | XP_014597075.1 | 224 | 84 | |||
| XM_023649837.1 | XP_023505605.1 | 1777 | 84 | |||
| XM_023649838.1 | XP_023505606.1 | 1192 | 84 | |||
| XM_023649839.1 | XP_023505607.1 | 1412 | 84 | |||
| XM_023649840.1 | XP_023505608.1 | 954 | 84 | |||
| XM_023649841.1 | XP_023505609.1 | 1205 | 84 | |||
| XM_023649843.1 | XP_023505611.1 | 1392 | 84 | |||
| XM_023649844.1 | XP_023505612.1 | 1388 | 84 | |||
| XM_023649845.1 | XP_023505613.1 | 1005 | 84 | |||
| XM_023649846.1 | XP_023505614.1 | 1124 | 84 | |||
| XM_023649847.1 | XP_023505615.1 | 1114 | 84 | |||
| XM_023649848.1 | XP_023505616.1 | 1307 | 84 | |||
| XM_023649849.1 | XP_023505617.1 | 502 | 84 | |||
| XM_023649850.1 | XP_023505618.1 | 316 | 84 | |||
| XM_023649852.1 | XP_023505620.1 | 498 | 84 | |||
| XM_023649853.1 | XP_023505621.1 | 312 | 84 | |||
| XM_023649854.1 | XP_023505622.1 | 1590 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014741399.2, XM_014741473.2, XM_014741528.2, XM_014741583.2, XM_014741589.2, XM_023649837.1, XM_023649838.1, XM_023649839.1, XM_023649840.1, XM_023649841.1, XM_023649843.1, XM_023649844.1, XM_023649845.1, XM_023649846.1, XM_023649847.1, XM_023649848.1, XM_023649849.1, XM_023649850.1, XM_023649852.1, XM_023649853.1, XM_023649854.1 transcripts.
Set Membership: |
|

ZNF45
Genomic Map