Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741399.2 | XP_014596885.1 | 1841 | 68 | ||
| XM_014741473.2 | XP_014596959.1 | 1379 | 68 | |||
| XM_014741528.2 | XP_014597014.1 | 1654 | 68 | |||
| XM_014741583.2 | XP_014597069.1 | 571 | 68 | |||
| XM_014741589.2 | XP_014597075.1 | 292 | 68 | |||
| XM_023649837.1 | XP_023505605.1 | 1845 | 68 | |||
| XM_023649838.1 | XP_023505606.1 | 1260 | 68 | |||
| XM_023649839.1 | XP_023505607.1 | 1480 | 68 | |||
| XM_023649840.1 | XP_023505608.1 | 1022 | 68 | |||
| XM_023649841.1 | XP_023505609.1 | 1273 | 68 | |||
| XM_023649843.1 | XP_023505611.1 | 1460 | 68 | |||
| XM_023649844.1 | XP_023505612.1 | 1456 | 68 | |||
| XM_023649845.1 | XP_023505613.1 | 1073 | 68 | |||
| XM_023649846.1 | XP_023505614.1 | 1192 | 68 | |||
| XM_023649847.1 | XP_023505615.1 | 1182 | 68 | |||
| XM_023649848.1 | XP_023505616.1 | 1375 | 68 | |||
| XM_023649849.1 | XP_023505617.1 | 570 | 68 | |||
| XM_023649850.1 | XP_023505618.1 | 384 | 68 | |||
| XM_023649852.1 | XP_023505620.1 | 566 | 68 | |||
| XM_023649853.1 | XP_023505621.1 | 380 | 68 | |||
| XM_023649854.1 | XP_023505622.1 | 1658 | 68 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014741399.2, XM_014741473.2, XM_014741528.2, XM_014741583.2, XM_014741589.2, XM_023649837.1, XM_023649838.1, XM_023649839.1, XM_023649840.1, XM_023649841.1, XM_023649843.1, XM_023649844.1, XM_023649845.1, XM_023649846.1, XM_023649847.1, XM_023649848.1, XM_023649849.1, XM_023649850.1, XM_023649852.1, XM_023649853.1, XM_023649854.1 transcripts.
Set Membership: |
|

ZNF45
Genomic Map