Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741399.2 | XP_014596885.1 | 2139 | 108 | ||
| XM_014741473.2 | XP_014596959.1 | 1677 | 108 | |||
| XM_014741528.2 | XP_014597014.1 | 1952 | 108 | |||
| XM_014741583.2 | XP_014597069.1 | 869 | 108 | |||
| XM_014741589.2 | XP_014597075.1 | 590 | 108 | |||
| XM_023649837.1 | XP_023505605.1 | 2143 | 108 | |||
| XM_023649838.1 | XP_023505606.1 | 1558 | 108 | |||
| XM_023649839.1 | XP_023505607.1 | 1778 | 108 | |||
| XM_023649840.1 | XP_023505608.1 | 1320 | 108 | |||
| XM_023649841.1 | XP_023505609.1 | 1571 | 108 | |||
| XM_023649843.1 | XP_023505611.1 | 1758 | 108 | |||
| XM_023649844.1 | XP_023505612.1 | 1754 | 108 | |||
| XM_023649845.1 | XP_023505613.1 | 1371 | 108 | |||
| XM_023649846.1 | XP_023505614.1 | 1490 | 108 | |||
| XM_023649847.1 | XP_023505615.1 | 1480 | 108 | |||
| XM_023649848.1 | XP_023505616.1 | 1673 | 108 | |||
| XM_023649849.1 | XP_023505617.1 | 868 | 108 | |||
| XM_023649850.1 | XP_023505618.1 | 682 | 108 | |||
| XM_023649852.1 | XP_023505620.1 | 864 | 108 | |||
| XM_023649853.1 | XP_023505621.1 | 678 | 108 | |||
| XM_023649854.1 | XP_023505622.1 | 1932 | 108 | |||
More Information
Set Membership: |
|

ZNF45
Genomic Map