Assay Details
| Gene Symbol: LRRC4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650233.1 | XP_023506001.1 | 594 | 107 | ||
| XM_023650234.1 | XP_023506002.1 | 416 | 107 | |||
More Information
Set Membership: |
|

LRRC4B
Genomic Map