Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023651502.1 | XP_023507270.1 | 157 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023651502.1 transcripts.
Set Membership: |
|

ZNF444
Genomic Map