Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023651495.1 | XP_023507263.1 | 721 | 100 | ||
| XM_023651496.1 | XP_023507264.1 | 542 | 100 | |||
| XM_023651497.1 | XP_023507265.1 | 506 | 100 | |||
| XM_023651498.1 | XP_023507266.1 | 2730 | 100 | |||
| XM_023651499.1 | XP_023507267.1 | 606 | 100 | |||
| XM_023651500.1 | XP_023507268.1 | 2099 | 100 | |||
| XM_023651501.1 | XP_023507269.1 | 483 | 100 | |||
| XM_023651502.1 | XP_023507270.1 | 516 | 100 | |||
More Information
Set Membership: |
|

ZNF444
Genomic Map