Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597105.2 | XP_005597162.1 | 671 | 65 | ||
XM_005597106.3 | XP_005597163.1 | 1258 | 65 | |||
XM_005597107.3 | XP_005597164.1 | 790 | 65 | |||
XM_023651940.1 | XP_023507708.1 | 1289 | 65 | |||
XM_023651941.1 | XP_023507709.1 | 973 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
